Merck
  • Metabolism of benzyl alcohol via catechol ortho-pathway in methylnaphthalene-degrading Pseudomonas putida CSV86.
Metabolism of benzyl alcohol via catechol ortho-pathway in methylnaphthalene-degrading Pseudomonas putida CSV86.
Applied microbiology and biotechnology (2003-04-11)
A Basu, S S Dixit, P S Phale
ABSTRACT
Pseudomonas putida CSV86 metabolizes 1- and 2-methylnaphthalene through distinct catabolic and detoxification pathways. In spite of the similarity in the steps involved in the methylnaphthalene detoxification and the toluene side-chain hydroxylation pathways, the strain failed to utilize toluene or xylenes. However, it could grow on benzyl alcohol, 2- and 4-hydroxybenzyl alcohol. Metabolic studies suggest that the benzyl alcohol metabolism proceeds via the benzaldehyde, benzoate, and catechol ortho-cleavage pathway, in contrast to the well established catechol meta-cleavage pathway. Carbon source-dependent enzyme activity studies suggest that the degradation of aromatic alcohol involves two regulons. Aromatic alcohol induces the upper regulon, which codes for aromatic alcohol- and aromatic aldehyde-dehydrogenase and converts alcohol into acid. The aromatic acid so generated induces the specific lower regulon and is metabolized via either the ortho- or the meta-cleavage pathway. CSV86 cells transform 1- and 2-methylnaphthalene to 1- and 2-hydroxymethyl naphthalene, which are further converted to the respective naphthoic acids due to the basal level expression and broad substrate specificity of the upper regulon enzymes.
Sigma-Aldrich
2-Methylnaphthalene (β), 95%
Supelco
2-Methylnaphthalene, analytical standard
Sigma-Aldrich
1-Methylnaphthalene, 95%
Sigma-Aldrich
1-Methylnaphthalene, ≥95%
Supelco
1-Methylnaphthalene, analytical standard