- Visualizing tmRNA entry into a stalled ribosome.
Visualizing tmRNA entry into a stalled ribosome.
Science (New York, N.Y.) (2003-04-05)
Mikel Valle, Reynald Gillet, Sukhjit Kaur, Anke Henne, V Ramakrishnan, Joachim Frank
PMID12677067
ABSTRACT
Bacterial ribosomes stalled on defective messenger RNAs (mRNAs) are rescued by tmRNA, an approximately 300-nucleotide-long molecule that functions as both transfer RNA (tRNA) and mRNA. Translation then switches from the defective message to a short open reading frame on tmRNA that tags the defective nascent peptide chain for degradation. However, the mechanism by which tmRNA can enter and move through the ribosome is unknown. We present a cryo-electron microscopy study at approximately 13 to 15 angstroms of the entry of tmRNA into the ribosome. The structure reveals how tmRNA could move through the ribosome despite its complicated topology and also suggests roles for proteins S1 and SmpB in the function of tmRNA.