Merck
  • Sulfo-SADP (sulfosuccinimidyl[4-azidophenyldithio]propionate) an active site directed reagent inhibiting the NADPH dependent O2- generation of leukocyte cytochrome b(558).
Sulfo-SADP (sulfosuccinimidyl[4-azidophenyldithio]propionate) an active site directed reagent inhibiting the NADPH dependent O2- generation of leukocyte cytochrome b(558).
Journal of biochemistry, molecular biology, and biophysics : JBMBB : the official journal of the Federation of Asian and Oceanian Biochemists and Molecular Biologists (FAOBMB) (2002-08-21)
Ming Cheng, Richard John Guillory
ABSTRACT
Functional reagents known to bring about the formation of a distinct membrane molecular complex of the subunits of cytochrome b(558) (gp 91(phox) and p22(phox)) were investigated for their influence on the O2- generating capability of liposome incorporated cytochrome b(558) preparations. One, ethyleneglycolbis[sulfo-succinimidylsuccinate], (sulfo-EGS) was found to inhibit O2- generation at concentrations which are known to result in cross-linking the two subunits of cytochrome b(558). Sulfosuccinimidyl [4-azidophenyldithio] propionate, (sulfo-SADP) on the other hand, was found to be a powerful inhibitor of the cytochrome b(558) dependent O2- production at concentrations not able to result in cross linking of the two subunits. Sulfo-SADP inhibits the cytochrome b(558) O2- production 50% at 25 microM, while sulfo-EGS requires 400 microM. For these reagents, the succinimidyl group of sulfo-SADP and sulfo-EGS is the reactive group, which inhibit irreversibly, cytochrome b(558) generation of O2-. Both sulfo-SADP and sulfo-EGS have similar linker arms of 13.9 and 16.1 A, respectively. The difference, accounting for the strong inhibitory profile for sulfo-SADP as compared with sulfo-EGS, resides in the aryl group associated with the sulfo-SADP. The aryl group of sulfo-SADP has been found to be important in directing the specificity of the probe in its inhibition of O2- generation. When the disulfide bond linking the aromatic portion of the probe to the succinimidyl ring is cleaved by DTT (dithiothreitol), the product loses its specificity and has an inhibitory activity with respect to O2- generation comparable to that of sulfo-EGS. The partial protection against the inhibitory influence of sulfo-SADP by NADP(+) indicates that the reagent may interact at the pyridine nucleotide-binding domain of cytochrome b(558). Its low inhibitory titer and its water solubility suggest that sulfo-SADP reacts with a specific amine (the primary reactant for the succinimidyl group) on cytochrome b(558).
Sigma-Aldrich
Ethylene glycol-bis(succinic acid N-hydroxysuccinimide ester), powder