Merck
  • Genotoxicity of trivalent chromium in bacterial cells. Possible effects on DNA topology.
Genotoxicity of trivalent chromium in bacterial cells. Possible effects on DNA topology.
Chemical research in toxicology (2002-07-18)
Andreja Plaper, Spela Jenko-Brinovec, Ales Premzl, Janko Kos, Peter Raspor
ABSTRACT
Trivalent chromium is a metal required for proper sugar and fat metabolism. However, it has been suggested that it causes DNA damage in in vitro test systems, although in vivo toxicity has not yet been proved. In the present study, the effect of Cr3+ on bacterial cells was tested with the Pro-Tox (C) assay, and its cellular uptake was measured with flame atomic absorption spectroscopy. The potential genotoxicity of Cr3+ was further examined by the study of its influence on a bacterial type II topoisomerase. Cr3+ was shown to cause DNA damage and inhibit topoisomerase DNA relaxation activity, probably by preventing the formation of the covalent link between enzyme and double helix. In addition, Cr3+ decreases the viability and/or proliferation rate of eukaryotic cells such as murine B16 melanoma cells and human MCF-10A neoT ras-transformed human epithelial cells. The possible implication for Cr3+ intake by humans is discussed.
Sigma-Aldrich
Chromium(III) chloride, purified by sublimation, 99%
Sigma-Aldrich
Chromium(III) chloride, anhydrous, 99.99% trace metals basis
Sigma-Aldrich
Chromium(III) nitrate nonahydrate, 99%
Sigma-Aldrich
Chromium(III) nitrate nonahydrate, ≥99.99% trace metals basis