Merck
  • Neutron structure and mechanistic studies of diisopropyl fluorophosphatase (DFPase).
Neutron structure and mechanistic studies of diisopropyl fluorophosphatase (DFPase).
Acta crystallographica. Section D, Biological crystallography (2010-11-03)
Julian C H Chen, Marat Mustyakimov, Benno P Schoenborn, Paul Langan, Marc Michael Blum
ABSTRACT
Diisopropyl fluorophosphatase (DFPase) is a calcium-dependent phosphotriesterase that acts on a variety of highly toxic organophosphorus compounds that act as inhibitors of acetylcholinesterase. The mechanism of DFPase has been probed using a variety of methods, including isotopic labelling, which demonstrated the presence of a phosphoenzyme intermediate in the reaction mechanism. In order to further elucidate the mechanism of DFPase and to ascertain the protonation states of the residues and solvent molecules in the active site, the neutron structure of DFPase was solved at 2.2 Å resolution. The proposed nucleophile Asp229 is deprotonated, while the active-site solvent molecule W33 was identified as water and not hydroxide. These data support a mechanism involving direct nucleophilic attack by Asp229 on the substrate and rule out a mechanism involving metal-assisted water activation. These data also allowed for the re-engineering of DFPase through rational design to bind and productively orient the more toxic S(P) stereoisomers of the nerve agents sarin and cyclosarin, creating a modified enzyme with enhanced overall activity and significantly increased detoxification properties.
Sigma-Aldrich
Diisopropyl-fluorophosphatase, ≥30 U/mg
Pricing and availability is not currently available.