Merck
  • Differential blockade of gamma-aminobutyric acid type A receptors by the neuroactive steroid dehydroepiandrosterone sulfate in posterior and intermediate pituitary.
Differential blockade of gamma-aminobutyric acid type A receptors by the neuroactive steroid dehydroepiandrosterone sulfate in posterior and intermediate pituitary.
Molecular pharmacology (1999-03-03)
S L Hansen, B Fjalland, M B Jackson
ABSTRACT
Dehydroepiandrosterone sulfate (DHEAS) is a neuroactive steroid with antagonist action at gamma-aminobutyric acid type A (GABAA) receptors. Patch-clamp techniques were used to investigate DHEAS actions at GABAA receptors of the rat pituitary gland at two distinct loci: posterior pituitary nerve terminals and intermediate pituitary endocrine cells. The GABA responses in these two regions were quite different, with posterior pituitary responses having smaller amplitudes and desensitizing more rapidly and more completely. DHEAS blockade of GABAA receptors in the two regions also was different. In posterior pituitary, a site with an apparent dissociation constant of 15 microM accounted for most of the blockade, but a small fraction of blockade may be related to a site with a dissociation constant in the nanomolar range. In the intermediate lobe, DHEAS sensitivities in the nanomolar and micromolar ranges were clearly evident, in proportions that varied widely from cell to cell. Regardless of whether the GABA response of a cell was highly sensitive or weakly sensitive to DHEAS, GABA alone evoked currents that were indistinguishable in terms of amplitude, desensitization kinetics, and GABA sensitivity. Thus, the structural elements responsible for DHEAS blockade have a highly selective impact on receptor function. GABAA receptors with nanomolar sensitivity to DHEAS have not been described previously. This suggests that DHEAS may have an important role in the modulation of neuropeptide secretion, and the diverse properties of GABAA receptors in the rat pituitary provide mechanisms for selective regulation of the different peptidergic systems of this gland.
Sigma-Aldrich
Dehydroisoandrosterone 3-sulfate sodium salt dihydrate, ≥93% (TLC), powder