Merck
  • Salivary cortisol, alpha-amylase and heart rate variation in response to dental treatment in children.
Salivary cortisol, alpha-amylase and heart rate variation in response to dental treatment in children.
The Journal of clinical pediatric dentistry (2013-01-25)
N F Furlan, M B D Gavião, T S Barbosa, J Nicolau, P M Castelo
ABSTRACT
Anxiety and stress are usually related to the dental treatment situation. The objective was to investigate salivary cortisol and alpha-amylase levels (salivary biomarkers) and heart rate in children undergoing a minor dental procedure (dental prophylaxis). In total, 31 children (range 84-95 months) of both genders without caries or history of dental treatment/pain/trauma were selected. Three saliva samples were gathered: one prior to dental prophylaxis, one immediately after and one ten minutes later. Weight and height were assessed, and heart rate was evaluated prior to and during the procedure. Data were analyzed by correlation tests and t-test/Wilcoxon (alpha = 0.05). Higher cortisol and amylase levels were observed before prophylaxis compared to afterward. Cortisol and amylase levels did not show a significant correlation, nor did salivary biomarkers and body mass index. However, heart rate and amylase levels showed a significant positive correlation. In the studied sample, certain anticipation of the dental treatment was observed because higher cortisol and amylase levels were observed before, rather than after the event; moreover a significant correlation between amylase levels and heart rate was observed Thus, salivary biomarkers may be a valuable tool for evaluating anxiety-producing events, such as dental treatment, in children.
Sigma-Aldrich
Taka-Diastase from Aspergillus oryzae, powder, slightly beige, ~100 U/mg
Sigma-Aldrich
α-Amylase from Bacillus sp., powder, yellow-brown, ~50 U/mg
Sigma-Aldrich
α-Amylase from Bacillus sp., powder, yellow-brown, ~380 U/mg
Sigma-Aldrich
α-Amylase from Bacillus sp., powder, ≥400 units/mg protein (Lowry)
Sigma-Aldrich
α-Amylase from porcine pancreas, PMSF Treated, Type I-A, saline suspension, ≥1000 units/mg protein (E1%/280)
Sigma-Aldrich
α-Amylase from porcine pancreas, Type I-A, PMSF treated, saline suspension, 700-1400 units/mg protein (E1%/280)
Sigma-Aldrich
α-Amylase, heat-stable, solution, for use in Total Dietary Fiber Assay, TDF-100A
Sigma-Aldrich
α-Amylase from porcine pancreas, Type VI-B, ≥5 units/mg solid
Sigma-Aldrich
α-Amylase from Bacillus sp., Type II-A, lyophilized powder, ≥1,500 units/mg protein (biuret)
Sigma-Aldrich
α-Amylase from Bacillus sp., liquid
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, aqueous solution, ≥800 FAU/g
Sigma-Aldrich
α-Amylase from Bacillus amyloliquefaciens, liquid, ≥250 units/g
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, ≥150 units/mg protein (biuret)
Sigma-Aldrich
α-Amylase from Bacillus licheniformis, Type XII-A, saline solution, ≥500 units/mg protein (biuret)
Supelco
α-Amylase from Bacillus licheniformis, suitable for determination of starch (Kit STA-20)
Sigma-Aldrich
α-Amylase from human saliva, Type XIII-A, lyophilized powder, 300-1,500 units/mg protein
Sigma-Aldrich
α-Amylase from human saliva, Type IX-A, lyophilized powder, 1,000-3,000 units/mg protein
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, powder, ~30 U/mg
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, powder, ~1.5 U/mg (~0.2 U acc. to Willstätter)
Sigma-Aldrich
α-Amylase from Bacillus licheniformis, lyophilized powder, 500-1,500 units/mg protein, 93-100% (SDS-PAGE)