Merck
  • Optimization of the pepsin digestion method for anisakids inspection in the fishing industry.
Optimization of the pepsin digestion method for anisakids inspection in the fishing industry.
Veterinary parasitology (2012-10-10)
María Llarena-Reino, Carmen Piñeiro, José Antonio, Luis Outeriño, Carlos Vello, Ángel F González, Santiago Pascual
ABSTRACT
During the last 50 years human anisakiasis has been rising while parasites have increased their prevalence at determined fisheries becoming an emergent major public health problem. Although artificial enzymatic digestion procedure by CODEX (STAN 244-2004: standard for salted Atlantic herring and salted sprat) is the recommended protocol for anisakids inspection, no international agreement has been achieved in veterinary and scientific digestion protocols to regulate this growing source of biological hazard in fish products. The aim of this work was to optimize the current artificial digestion protocol by CODEX with the purpose of offering a faster, more useful and safer procedure for factories workers, than the current one for anisakids detection. To achieve these objectives, the existing pepsin chemicals and the conditions of the digestion method were evaluated and assayed in fresh and frozen samples, both in lean and fatty fish species. Results showed that the new digestion procedure considerably reduces the assay time, and it is more handy and efficient (the quantity of the resulting residue was considerably lower after less time) than the widely used CODEX procedure. In conclusion, the new digestion method herein proposed based on liquid pepsin format is an accurate reproducible and user-friendly off-site tool, that can be useful in the implementation of screening programs for the prevention of human anisakiasis (and associated gastroallergic disorders) due to the consumption of raw or undercooked contaminated seafood products.
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, slightly beige, 1200-2400 U/mg
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, slightly beige, ≥500 U/mg
Sigma-Aldrich
Pepsin from porcine gastric mucosa, tested according to Ph. Eur.
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, ≥400 units/mg protein
Sigma-Aldrich
Pepsin from porcine gastric mucosa, lyophilized powder, ≥3,200 units/mg protein
Sigma-Aldrich
Pepsin−Agarose from porcine gastric mucosa, lyophilized powder, ≥30 units/mg dry solid
Sigma-Aldrich
Pepsin from porcine gastric mucosa, lyophilized powder, ≥2,500 units/mg protein (E1%/280)
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, ≥250 units/mg solid