Merck
  • Construction of the intermediate vector pVBG2307 by incorporating vital elements of expression vectors pBI121 and pBI221.
Construction of the intermediate vector pVBG2307 by incorporating vital elements of expression vectors pBI121 and pBI221.
Genetics and molecular research : GMR (2012-09-26)
S S Ahmed, Z-H Gong, J-J Ji, Y-X Yin, H-J Xiao, M A Khan, A Rehman, I Ahmad
ABSTRACT
Molecular chaperones of plasmid pBI121 carrying CaMV35S promoter and a nucleotide sequence of plasmid pBI221 were inserted into plasmid pCAMBIA2300 to construct an intermediate vector: pVBG2307. This novel vector pVBG2307 contains a greatly expanded multiple cloning site with an adjacent imported CaMV35S promoter sequence. This vector allows controlled transformation of DNA in both Escherichia coli and Agrobacterium tumefaciens. Cloned PG, orf456, ipt genes and E8, a fruiting promoter, were amplified by PCR of cDNA libraries of Capsicum annum and Lycopersicon esculentum and were then transferred into vector pVBG2307. The viability of this vector was demonstrated, as it regulated PG, orf456, ipt and E8 genes in E. coli and could be transferred into Agrobacterium strain EHA105-4.
Sigma-Aldrich
Pectinase from Aspergillus niger, powder, slightly beige, >1 U/mg
Sigma-Aldrich
Pectinase from Aspergillus niger, BioReagent, suitable for plant cell culture, aqueous glycerol solution, ≥5 units/mg protein (Lowry)
Sigma-Aldrich
Pectinase from Rhizopus sp., powder, 400-800 units/g solid
Sigma-Aldrich
Pectolyase from Aspergillus japonicus, lyophilized powder, ≥0.3 units/mg solid
Sigma-Aldrich
Pectolyase from Aspergillus japonicus, suitable for plant cell culture, lyophilized powder