Merck
  • Immobilization of pectinase and lipase on macroporous resin coated with chitosan for treatment of whitewater from papermaking.
Immobilization of pectinase and lipase on macroporous resin coated with chitosan for treatment of whitewater from papermaking.
Bioresource technology (2012-09-06)
Kai Liu, Guanglei Zhao, Beihai He, Lihui Chen, Liulian Huang
ABSTRACT
Anionic residues and pitch deposits in whitewater negatively impact the operation of paper-forming equipment. In order to remove these substances, a macroporous resin based on a methyl acrylate matrix was synthesized and coated with chitosan of various molecular weights through glutaraldehyde cross-linking. Pectinase from Bacillus licheniformis and lipase from Thermomyces lanuginosus were immobilized on the resin coated with chitosan by a Schiff base reaction. The highest hydrolysis activities of the immobilized enzymes were achieved by using chitosan with 10×10(5)DaMW for coating and 0.0025% glutaraldehyde for cross-linking chitosan. The cationic demand and pitch deposits in whitewater were reduced by 58% and 74%, respectively, when treating whitewater with immobilized dual-enzymes for 15min at 55°C and pH 7.5. This method is useful for treatment of whitewater in the papermaking industry.
Sigma-Aldrich
Pectinase from Aspergillus niger, powder, slightly beige, >1 U/mg
Sigma-Aldrich
Pectinase from Aspergillus niger, BioReagent, suitable for plant cell culture, aqueous glycerol solution, ≥5 units/mg protein (Lowry)
Sigma-Aldrich
Pectolyase from Aspergillus japonicus, suitable for plant cell culture, lyophilized powder
Sigma-Aldrich
Pectinase from Rhizopus sp., powder, 400-800 units/g solid
Sigma-Aldrich
Pectolyase from Aspergillus japonicus, lyophilized powder, ≥0.3 units/mg solid