Merck
  • Construction of membrane-anchoring fusion protein of Thermococcus kodakaraensis glycerol kinase and its application to repetitive batchwise reactions.
Construction of membrane-anchoring fusion protein of Thermococcus kodakaraensis glycerol kinase and its application to repetitive batchwise reactions.
Journal of bioscience and bioengineering (2011-12-20)
Elvi Restiawaty, Kohsuke Honda, Kenji Okano, Ryuichi Hirota, Takeshi Omasa, Akio Kuroda, Hisao Ohtake
ABSTRACT
We previously demonstrated the stoichiometric conversion of glycerol to glycerol-3-phosphate (G3P) using Escherichia coli recombinants producing the ATP-dependent glycerol kinase of the hyperthermophile Thermococcus kodakaraensis (TkGK) and the polyphosphate kinase of Thermus thermophilus HB27 (TtPPK). TtPPK was associated with the membrane fraction of E. coli recombinants, whereas TkGK was released from the cells during the reaction at 70°C. In this study, TkGK was fused with either TtPPK or an E. coli membrane-intrinsic protein, YedZ, to minimize the heat-induced leakage of TkGK. When the E. coli recombinants having these fusion proteins were incubated at 70°C for 2h, more than 80% of TkGK activity was retained in the heated E. coli cells. However, the yields of G3P production by E. coli having the fusion proteins of TtPPK and TkGK were only less than 35%. Polyphosphate is a strong chelator for metal ions and has an inhibitory effect on TkGK which requires magnesium. Insufficient space between TtPPK and TkGK might enhance the inhibitory effect of polyphosphate on TkGK activity of the fusion protein. The mixture of E. coli cells having TtPPK and those having TkGK fused with YedZ converted 80% of glycerol into G3P. These recombinant cells could be easily recovered from the reaction mixture by centrifugation and repeatedly used without a significant loss of enzyme activities.
Sigma-Aldrich
Glycerokinase from Bacillus stearothermophilus, buffered aqueous solution, ≥75 units/mg protein (biuret)
Sigma-Aldrich
Glycerokinase from Escherichia coli, 300-600 units/mL, ammonium sulfate suspension
Sigma-Aldrich
Glycerokinase from Escherichia coli, lyophilized powder, 40-100 units/mg protein
Sigma-Aldrich
Glycerokinase from Cellulomonas sp., lyophilized powder, 25-75 units/mg protein