Merck
Inhibitors of the bacterial cell wall biosynthesis enzyme MurD.
Bioorganic & medicinal chemistry letters (1999-01-05)
L D Gegnas, S T Waddell, R M Chabin, S Reddy, K K Wong
ABSTRACT
A series of transition-state analog inhibitors of the D-glutamic acid-adding enzyme (MurD) of bacterial peptidoglycan biosynthesis has been synthesized and evaluated for inhibition of the E. coli enzyme.
Sigma-Aldrich
5-Phospho-D-ribose 1-diphosphate pentasodium salt, ≥75% (HPLC)