Merck
Characterization of trans-neuronal trafficking of Cbln1.
Molecular and cellular neurosciences (2009-04-07)
Peng Wei, Yongqi Rong, Leyi Li, Dashi Bao, James I Morgan
ABSTRACT
Cbln1, a glycoprotein secreted from granule cells and GluRdelta2 in the postsynaptic densities of Purkinje cells are components of an incompletely understood pathway essential for integrity and plasticity of parallel fiber-Purkinje cell synapses. We show that Cbln1 undergoes anterograde transport from granule cells to Purkinje cells and Bergmann glia, and enters the endolysosomal trafficking system, raising the possibility that Cbln1 exerts its activity on or within Purkinje cells and Bergmann glia. Cbln1 is absent in Purkinje cells and Bergmann glia of GluRdelta2-null mice, suggesting a mechanistic convergence on Cbln1 trafficking. Ectopic expression of Cbln1 in Purkinje cells of L7-cbln1 transgenic mice reveals Cbln1 undergoes anterograde and retrograde trans-neuronal trafficking even across synapses that lack GluRDelta2, indicating that it is not universally essential for Cbln1 transport. The L7-cbln1 transgene also ameliorates the locomotor deficits of cbln1-null mice, indicating that the presence and/or release of Cbln1 from the postsynaptic neuron has functional consequences.
Sigma-Aldrich
TWEEN® 20, viscous liquid, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
TWEEN® 20, Low-peroxide; Low-carbonyls
Pricing and availability is not currently available.
Sigma-Aldrich
TWEEN® 20, Low-peroxide; Low-carbonyls
Pricing and availability is not currently available.
Sigma-Aldrich
TWEEN® 20, BioXtra, viscous liquid
Pricing and availability is not currently available.
Sigma-Aldrich
TWEEN® 20, viscous liquid
Pricing and availability is not currently available.
Sigma-Aldrich
TWEEN® 20, for molecular biology, viscous liquid
Pricing and availability is not currently available.
Sigma-Aldrich
TWEEN® 20, viscosity 250-450 mPa.s (25 °C)
Pricing and availability is not currently available.
Sigma-Aldrich
ECO TWEEN® 20, viscous liquid
Pricing and availability is not currently available.