Merck
  • FOXO-regulated OSER1 reduces oxidative stress and extends lifespan in multiple species.
FOXO-regulated OSER1 reduces oxidative stress and extends lifespan in multiple species.
Nature communications (2024-08-21)
Jiangbo Song, Zhiquan Li, Lei Zhou, Xin Chen, Wei Qi Guinevere Sew, Héctor Herranz, Zilu Ye, Jesper Velgaard Olsen, Yuan Li, Marianne Nygaard, Kaare Christensen, Xiaoling Tong, Vilhelm A Bohr, Lene Juel Rasmussen, Fangyin Dai
ABSTRACT
FOXO transcription factors modulate aging-related pathways and influence longevity in multiple species, but the transcriptional targets that mediate these effects remain largely unknown. Here, we identify an evolutionarily conserved FOXO target gene, Oxidative stress-responsive serine-rich protein 1 (OSER1), whose overexpression extends lifespan in silkworms, nematodes, and flies, while its depletion correspondingly shortens lifespan. In flies, overexpression of OSER1 increases resistance to oxidative stress, starvation, and heat shock, while OSER1-depleted flies are more vulnerable to these stressors. In silkworms, hydrogen peroxide both induces and is scavenged by OSER1 in vitro and in vivo. Knockdown of OSER1 in Caenorhabditis elegans leads to increased ROS production and shorter lifespan, mitochondrial fragmentation, decreased ATP production, and altered transcription of mitochondrial genes. Human proteomic analysis suggests that OSER1 plays roles in oxidative stress response, cellular senescence, and reproduction, which is consistent with the data and suggests that OSER1 could play a role in fertility in silkworms and nematodes. Human studies demonstrate that polymorphic variants in OSER1 are associated with human longevity. In summary, OSER1 is an evolutionarily conserved FOXO-regulated protein that improves resistance to oxidative stress, maintains mitochondrial functional integrity, and increases lifespan in multiple species. Additional studies will clarify the role of OSER1 as a critical effector of healthy aging.
Sigma-Aldrich
Anti-OSER1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution