Merck
  • Carbon monoxide blocks lipopolysaccharide-induced gene expression by interfering with proximal TLR4 to NF-kappaB signal transduction in human monocytes.
Carbon monoxide blocks lipopolysaccharide-induced gene expression by interfering with proximal TLR4 to NF-kappaB signal transduction in human monocytes.
PloS one (2009-12-04)
Maneesha Chhikara, Shuibang Wang, Steven J Kern, Gabriela A Ferreyra, Jennifer J Barb, Peter J Munson, Robert L Danner
ABSTRACT
Carbon monoxide (CO) is an endogenous messenger that suppresses inflammation, modulates apoptosis and promotes vascular remodeling. Here, microarrays were employed to globally characterize the CO (250 ppm) suppression of early (1 h) LPS-induced inflammation in human monocytic THP-1 cells. CO suppressed 79 of 101 immediate-early genes induced by LPS; 19% (15/79) were transcription factors and most others were cytokines, chemokines and immune response genes. The prototypic effects of CO on transcription and protein production occurred early but decreased rapidly. CO activated p38 MAPK, ERK1/2 and Akt and caused an early and transitory delay in LPS-induced JNK activation. However, selective inhibitors of these kinases failed to block CO suppression of LPS-induced IL-1beta, an inflammation marker. Of CO-suppressed genes, 81% (64/79) were found to have promoters with putative NF-kappaB binding sites. CO was subsequently shown to block LPS-induced phosphorylation and degradation of IkappaBalpha in human monocytes, thereby inhibiting NF-kappaB signal transduction. CO broadly suppresses the initial inflammatory response of human monocytes to LPS by reshaping proximal events in TLR4 signal transduction such as stress kinase responses and early NF-kappaB activation. These rapid, but transient effects of CO may have therapeutic applications in acute pulmonary and vascular injury.
Sigma-Aldrich
NuCLEAR Extraction Kit, For mammalian tissue or cultured cells
Sigma-Aldrich
2-Mercaptoethanol, for molecular biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
Nuclei Isolation Kit: Nuclei EZ Prep, sufficient for 25 nuclei preparations (~1-10×107 cells/preparation)
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Nuclei Isolation Kit: Nuclei PURE Prep, sufficient for 15 nuclei preparations (~1-10×107 cells or 1g of tissue per preparation)
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%