- Electron Microscopy of Cryptococcus neoformans: Processing Challenges to Avoid Artifacts.
Electron Microscopy of Cryptococcus neoformans: Processing Challenges to Avoid Artifacts.
Methods in molecular biology (Clifton, N.J.) (2024-05-17)
Glauber R de S Araújo, Bruno Pontes, Susana Frases
PMID38758316
ABSTRACT
This chapter describes methodological details for preparing specimens of Cryptococcus neoformans (although it can be applied to any species of the genus) and their subsequent analysis by scanning and transmission electron microscopy. Adaptations to conventional protocols for better preservation of the sample, as well as to avoid artifacts, are presented. The protocols may be used to examine both the surface ultrastructure and the interior of this pathogenic fungus in detail.