Merck
  • Non-apical mitoses contribute to cell delamination during mouse gastrulation.
Non-apical mitoses contribute to cell delamination during mouse gastrulation.
Nature communications (2024-08-31)
Evangéline Despin-Guitard, Viviane S Rosa, Steffen Plunder, Navrita Mathiah, Kristof Van Schoor, Eliana Nehme, Sara Merino-Aceituno, Joaquim Egea, Marta N Shahbazi, Eric Theveneau, Isabelle Migeotte
ABSTRACT
During the epithelial-mesenchymal transition driving mouse embryo gastrulation, cells divide more frequently at the primitive streak, and half of those divisions happen away from the apical pole. These observations suggest that non-apical mitoses might play a role in cell delamination. We aim to uncover and challenge the molecular determinants of mitosis position in different regions of the epiblast through computational modeling and pharmacological treatments of embryos and stem cell-based epiblast spheroids. Blocking basement membrane degradation at the streak has no impact on the asymmetry in mitosis frequency and position. By contrast, disturbance of the actomyosin cytoskeleton or cell cycle dynamics elicits ectopic non-apical mitosis and shows that the streak region is characterized by local relaxation of the actomyosin cytoskeleton and less stringent regulation of cell division. These factors are essential for normal dynamics at the streak and favor cell delamination from the epiblast.
Sigma-Aldrich
Anti-phospho-Histone H3.1 (pSer10) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Cdk1 Inhibitor IV, RO-3306, RO-3306 is a cell-permeable, potent and ATP-competitive inhibitor of Cdk1 (Ki = 35 nM and 110 nM for Cdk1/B1 and Cdk1/A, respectively).
Sigma-Aldrich
Anti-phospho-Histone H3 (Ser10) Antibody, Mitosis Marker, Upstate®, from rabbit
Sigma-Aldrich
Anti-Collagen Type IV Antibody, Chemicon®, from goat
Sigma-Aldrich
XAV939, ≥98% (HPLC)