Merck
  • Generation of 3 patient induced Pluripotent stem cell lines containing SORD mutations linked to a recessive neuropathy.
Generation of 3 patient induced Pluripotent stem cell lines containing SORD mutations linked to a recessive neuropathy.
Stem cell research (2024-05-27)
Christopher Yanick, Renata Maciel, Elizabeth Jacobs, Jacquelyn Schatzman, Michael Shy, Stephan Zuchner, Mario Saporta
ABSTRACT
The SORD neuropathy has been identified as the most common autosomal recessive inherited neuropathy, occurring in thousands of patients worldwide. Fibroblast lines from 3 different patients containing the c.753delG; p.Ala253GlnfsTer27 SORD mutations were reprogrammed into induced Pluripotent Stem Cell (iPSC) lines. These iPSC lines demonstrate an apparent normal karyotype and have positive expression of pluripotency markers. These iPSC lines also stain positively for Ectoderm, Endoderm and Mesoderm markers following Embryoid body differentiation. These lines pose to serve as a valuable disease modeling resource for studying the SORD neuropathy, including studying disease phenotype and treatment efficacy.
Sigma-Aldrich
Anti-Sox2 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-OCT-4 [POU5F1] Antibody, clone 7F9.2, Alexa Fluor 488 conjugate, clone 7F9.2, from mouse, ALEXA FLUOR 488
Sigma-Aldrich
Anti-NANOG Antibody, clone 7F7.1, Alexa Fluor 488 conjugate, clone 7F7.1, from mouse, ALEXA FLUOR 488
Sigma-Aldrich
Anti-β-Tubulin III antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution