Merck
  • Predicting Atrophy of the Cochlear Stria Vascularis from the Shape of the Threshold Audiogram.
Predicting Atrophy of the Cochlear Stria Vascularis from the Shape of the Threshold Audiogram.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2023-10-21)
Charanjeet Kaur, Pei-Zhe Wu, Jennifer T O'Malley, M Charles Liberman
ABSTRACT
Several lines of evidence have suggested that steeply sloping audiometric losses are caused by hair cell degeneration, while flat audiometric losses are caused by strial atrophy, but this concept has never been rigorously tested in human specimens. Here, we systematically compare audiograms and cochlear histopathology in 160 human cases from the archival collection of celloidin-embedded temporal bones at the Massachusetts Eye and Ear. The dataset included 106 cases from a prior study of normal-aging ears, and an additional 54 cases selected by combing the database for flat audiograms. Audiogram shapes were classified algorithmically into five groups according to the relation between flatness (i.e., SD of hearing levels across all frequencies) and low-frequency pure-tone average (i.e., mean at 0.25, 0.5, and 1.0 kHz). Outer and inner hair cell losses, neural degeneration, and strial atrophy were all quantified as a function of cochlear location in each case. Results showed that strial atrophy was worse in the apical than the basal half of the cochlea and was worse in females than in males. The degree of strial atrophy was uncorrelated with audiogram flatness. Apical atrophy was correlated with low-frequency thresholds and basal atrophy with high-frequency thresholds, and the former correlation was higher. However, a multivariable regression with all histopathological measures as predictors and audiometric thresholds as the outcome showed that strial atrophy was a significant predictor of threshold shift only in the low-frequency region, and, even there, the contribution of outer hair cell damage was larger.SIGNIFICANCE STATEMENT Cochlear pathology can only be assessed postmortem; thus, human cochlear histopathology is critical to our understanding of the mechanisms of hearing loss. Dogma holds that relative damage to sensory cells, which transduce mechanical vibration into electrical signals, versus the stria vascularis, the cellular battery that powers transduction, can be inferred by the shape of the audiogram, that is, down-sloping (hair cell damage) versus flat (strial atrophy). Here we quantified hair cell and strial atrophy in 160 human specimens to show that it is the degree of low-frequency hearing loss, rather than the audiogram slope, that predicts strial atrophy. Results are critical to the design of clinical trials for hearing-loss therapeutics, as current drugs target only hair cell, not strial, regeneration.
Sigma-Aldrich
Anti-Cochlin Antibody/COCH, clone 9A10D2, clone 9A10D2, from rat