Merck
  • Role and mechanism of PVN-sympathetic-adipose circuit in depression and insulin resistance induced by chronic stress.
Role and mechanism of PVN-sympathetic-adipose circuit in depression and insulin resistance induced by chronic stress.
EMBO reports (2023-10-23)
Jing Wang, Linshan Sun, Jingjing You, Honghai Peng, Haijing Yan, Jiangong Wang, Fengjiao Sun, Minghu Cui, Sanwang Wang, Zheng Zhang, Xueli Fan, Dunjiang Liu, Cuilan Liu, Changyun Qiu, Chao Chen, Zhicheng Xu, Jinbo Chen, Wei Li, Bin Liu
ABSTRACT
Chronic stress induces depression and insulin resistance, between which there is a bidirectional relationship. However, the mechanisms underlying this comorbidity remain unclear. White adipose tissue (WAT), innervated by sympathetic nerves, serves as a central node in the interorgan crosstalk through adipokines. Abnormal secretion of adipokines is involved in mood disorders and metabolic morbidities. We describe here a brain-sympathetic nerve-adipose circuit originating in the hypothalamic paraventricular nucleus (PVN) with a role in depression and insulin resistance induced by chronic stress. PVN neurons are labelled after inoculation of pseudorabies virus (PRV) into WAT and are activated under restraint stress. Chemogenetic manipulations suggest a role for the PVN in depression and insulin resistance. Chronic stress increases the sympathetic innervation of WAT and downregulates several antidepressant and insulin-sensitizing adipokines, including leptin, adiponectin, Angptl4 and Sfrp5. Chronic activation of the PVN has similar effects. β-adrenergic receptors translate sympathetic tone into an adipose response, inducing downregulation of those adipokines and depressive-like behaviours and insulin resistance. We finally show that AP-1 has a role in the regulation of adipokine expression under chronic stress.
Sigma-Aldrich
Anti-JUND antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-FRA-2 Antibody, clone REY146C, clone REY146C, from rat
Sigma-Aldrich
Anti-Tyrosine Hydroxylase Antibody, Chemicon®, from rabbit