Merck
Microscopy as a diagnostic tool in pulmonary tuberculosis.
International journal of mycobacteriology (2015-12-15)
Ritu Singhal, Vithal Prasad Myneedu
ABSTRACT
Tuberculosis continues to cast a huge impact on humanity with its high incidence and mortality, especially in developing countries. For tuberculosis case detection, microscopy continues to be indispensible, given its low cost, rapidity, simplicity of procedure and high specificity. Modifications have attempted to improve the sensitivity of microscopy which include: concentration methods such as centrifugation, N-acetyl cysteine-sodium hydroxide, bleach, ammonium sulfate or chitin. Furthermore, classical Ziehl-Neelsen (ZN) staining has been subjected to varying carbol fuchsin concentrations or replaced by Kinyoun staining, fluorescent microscopy or immune-fluorescence. Currently, light emitting diode fluorescence is recognizably the most plausible method as an alternative to ZN staining.
Sigma-Aldrich
Carbol-Fuchsin solution according to Kinyoun