Merck
  • Circadian protein TIMELESS regulates synaptic function and memory by modulating cAMP signaling.
Circadian protein TIMELESS regulates synaptic function and memory by modulating cAMP signaling.
Cell reports (2023-04-13)
Estibaliz Barrio-Alonso, Pablo J Lituma, Michael J Notaras, Robert Albero, Youcef Bouchekioua, Natalie Wayland, Isidora N Stankovic, Tanya Jain, Sijia Gao, Diany Paola Calderon, Pablo E Castillo, Dilek Colak
ABSTRACT
The regulation of neurons by circadian clock genes is thought to contribute to the maintenance of neuronal functions that ultimately underlie animal behavior. However, the impact of specific circadian genes on cellular and molecular mechanisms controlling synaptic plasticity and cognitive function remains elusive. Here, we show that the expression of the circadian protein TIMELESS displays circadian rhythmicity in the mammalian hippocampus. We identify TIMELESS as a chromatin-bound protein that targets synaptic-plasticity-related genes such as phosphodiesterase 4B (Pde4b). By promoting Pde4b transcription, TIMELESS negatively regulates cAMP signaling to modulate AMPA receptor GluA1 function and influence synaptic plasticity. Conditional deletion of Timeless in the adult forebrain impairs working and contextual fear memory in mice. These cognitive phenotypes were accompanied by attenuation of hippocampal Schaffer-collateral synapse long-term potentiation. Together, these data establish a neuron-specific function of mammalian TIMELESS by defining a mechanism that regulates synaptic plasticity and cognitive function.
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
Picrotoxin, powder
Sigma-Aldrich
Anti-GluR1 Antibody, clone C3T, rabbit monoclonal, culture supernatant, clone C3T, Upstate®
Millipore
Protease Inhibitor Cocktail Set III, EDTA-Free, Protease inhibitor cocktail III, EDTA-free for inhibiting aspartic, cysteine, and serine proteases as well as aminopeptidases in mammalian cells and tissues.
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Monoclonal Anti-CaM Kinase IIα (CaMKIIα) antibody produced in mouse, clone 6G9, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-Glutamate Receptor 1 Antibody, phosphoSer 845, Chemicon®, from rabbit
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse