Merck
  • KLHL7 promotes hepatocellular carcinoma progression and molecular therapy resistance by degrading RASA2.
KLHL7 promotes hepatocellular carcinoma progression and molecular therapy resistance by degrading RASA2.
iScience (2023-06-28)
Lin Chen, Yun Li, Yongheng Chen
ABSTRACT
Hepatocellular carcinoma (HCC) is a common aggressive tumor with a poor prognosis, and patients often seem to be refractory to the use of therapeutic drugs. In this study, we found that the KLHL7 expression was upregulated in HCC that was associated with poor patient prognosis. KLHL7 has been found to promote HCC development in both in vitro and in vivo experiments. Mechanistically, RASA2, a RAS GAP, was identified as a substrate of KLHL7. Upregulation of KLHL7 by growth factors promotes K48-linked polyubiquitination of RASA2 for degradation via the proteasomal pathway. Our in vivo experiments revealed that inhibition of KLHL7 in combination with lenvatinib treatment resulted in efficient killing of HCC cells. Together, these findings demonstrate a role for KLHL7 in HCC and reveal a mechanism by which growth factors regulate the RAS-MAPK pathway. It represents a potential therapeutic target for HCC.
Sigma-Aldrich
Anti-KLHL7 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab1
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2-FITC antibody produced in mouse, clone M2, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)