Merck
  • Internalization of apoptotic cells during efferocytosis requires Mertk-mediated calcium influx.
Internalization of apoptotic cells during efferocytosis requires Mertk-mediated calcium influx.
Cell death & disease (2023-07-01)
Susumin Yang, Chanhyuk Min, Hyunji Moon, Byeongjin Moon, Juyeon Lee, Jaeseon Jeon, Hagyeong Kwon, Deokyun Jang, Daeho Park
ABSTRACT
Phagocytosis of apoptotic cells, called efferocytosis, requires calcium inside and outside of phagocytes. Due to its necessity, calcium flux is sophisticatedly modulated, and the level of intracellular calcium in phagocytes is ultimately elevated during efferocytosis. However, the role of elevated intracellular calcium in efferocytosis remains elusive. Here, we report that Mertk-mediated intracellular calcium elevation is necessary for internalization of apoptotic cells during efferocytosis. Drastic depletion of intracellular calcium abrogated the internalization step of efferocytosis by delaying phagocytic cup extension and closure. Especially, the defect of phagocytic cup closure for internalization of apoptotic cells was caused by impaired F-actin disassembly and the attenuated interaction of Calmodulin with myosin light chain kinase (MLCK), leading to diminished myosin light chain (MLC) phosphorylation. Genetic and pharmacological impairment of the Calmodulin-MLCK-MLC axis or Mertk-mediated calcium influx also resulted in inefficient efferocytosis due to a defect in internalization of the targets. Taken together, our observations imply that intracellular calcium elevation through Mertk-mediated calcium influx facilitates efferocytosis by inducing myosin II-mediated contraction and F-actin disassembly required for internalization of apoptotic cells.
Sigma-Aldrich
Anti-Rac1 Antibody, clone 23A8, clone 23A8, Upstate®, from mouse
Sigma-Aldrich
U-73122, U-73122, CAS 112648-68-7, inhibits agonist-induced phospholipase C activation (IC50 = 1-2.1 µM) in human platelets and neutrophils.
Sigma-Aldrich
Monoclonal Anti-Myosin Light Chain Kinase antibody produced in mouse, clone K36, ascites fluid
Sigma-Aldrich
W-7, Hydrochloride, A cell-permeable and reversible calmodulin antagonist that inhibits myosin light chain kinase (IC₅₀ = 51 µM) and Ca2+-calmodulin-dependent phosphodiesterase (IC₅₀ = 28 µM).
Sigma-Aldrich
ML-7, powder
Sigma-Aldrich
Anti-Calmodulin Antibody, Upstate®, from mouse