Merck
  • The Clinicopathological Significance of BiP/GRP-78 in Breast Cancer: A Meta-Analysis of Public Datasets and Immunohistochemical Detection.
The Clinicopathological Significance of BiP/GRP-78 in Breast Cancer: A Meta-Analysis of Public Datasets and Immunohistochemical Detection.
Current oncology (Toronto, Ont.) (2022-12-23)
Inês Direito, Daniela Gomes, Fátima Liliana Monteiro, Isa Carneiro, João Lobo, Rui Henrique, Carmen Jerónimo, Luisa Alejandra Helguero
ABSTRACT
The endoplasmic reticulum chaperone BiP (also known as GRP-78 or HSPA5) maintains protein folding to allow cell proliferation and survival and has been implicated in carcinogenesis, tumor progression, and therapy resistance. BiP's association with clinical factors and prognostic potential in breast cancer remains unclear. In this work, three types of analysis were conducted to improve the knowledge of BiP's clinicopathological potential: (1) analysis of publicly available RNA-seq and proteomics datasets stratified as high and low quartiles; (2) a systematic review and meta-analysis of immunohistochemical detection of BIP; (3) confirmation of findings by BiP immunohistochemical detection in two luminal-like breast cancer small cohorts of paired samples (pre- vs. post-endocrine therapy, and primary pre- vs. metastasis post-endocrine therapy). The TCGA PanCancer dataset and CPTAC showed groups with high BiP mRNA and protein associated with HER2, basal-like subtypes, and higher immune scores. The meta-analysis of BiP immunohistochemistry disclosed an association between higher BiP positivity and reduced relapse-free survival. BiP immunohistochemistry confirmed increased BiP expression in metastasis, an association of BiP positivity with HER2 expression, and nuclear BiP localization with higher a tumor stage and poor outcome. Therefore, three independent approaches showed that BiP protein is associated with worse outcomes and holds prognostic potential for breast cancer.
Sigma-Aldrich
Anti-Rabbit IgG (Fc specific)-Biotin antibody produced in goat, affinity isolated antibody, lyophilized powder
Sigma-Aldrich
Anti-HSPA5 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution