Merck
  • Clinical significance of FBXO43 in hepatocellular carcinoma and its impact on tumor cell proliferation, migration and invasion.
Clinical significance of FBXO43 in hepatocellular carcinoma and its impact on tumor cell proliferation, migration and invasion.
PeerJ (2023-05-30)
Rulan Ma, Wenbo Liu, Tuanhe Sun, Chengxue Dang, Kang Li
ABSTRACT
The effects of FBXO43 on hepatocellular carcinoma (HCC) and its clinical significance have not yet been determined. This study aims to determine the clinical significance of FBXO43 in HCC and its impact on the biological functions of HCC cells. Data from TCGA database were downloaded to investigate the expression of FBXO43 in HCC and its correlation with prognosis and immune infiltration. Immunohistochemical staining images of FBXO43 in HCC were acquired from the HPA website. HCC cells (BEL-7404 and SMMC-7721) were transfected with the lentivirus targeting FBXO43 to decrease FBXO43 expression in HCC cells. Western blotting assay was conducted to evaluate the expression level of FBXO43 protein. MTT assay was used to detect the proliferation of HCC cells. The migration and invasion of HCC cells were investigated by performing scratch wound-healing and Transwell invasion assays, respectively. In comparison to normal tissues, FBXO43 is overexpressed in HCC tissue, and high FBXO43 expression is linked to late T stage, TNM stage and tumor grade. Elevated FBXO43 expression is a risk factor for HCC. In patients with high FBXO43 expression, the overall survival, disease-specific survival, progression-free survival and disease-free survival are poorer. The proliferation, migration and invasion of HCC cells are significantly attenuated in FBXO43 knockdown cells. Also, TCGA data analysis reveals that FBXO43 exhibits a positive correlation with immunosuppression of HCC. FBXO43 is overexpressed in HCC, and is linked to late tumor stage, worse prognosis and tumor immunosuppression. FBXO43 knockdown restrains the proliferation, migration and invasion of HCC.
Sigma-Aldrich
Anti-FBXO43 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1