Merck
  • Metastatic pancreatic neuroendocrine tumors feature elevated T cell infiltration.
Metastatic pancreatic neuroendocrine tumors feature elevated T cell infiltration.
JCI insight (2022-10-28)
Jacques Greenberg, Jessica Limberg, Akanksha Verma, David Kim, Xiang Chen, Yeon J Lee, Maureen D Moore, Timothy M Ullmann, Jessica W Thiesmeyer, Zachary Loewenstein, Kevin J Chen, Caitlin E Egan, Dessislava Stefanova, Rohan Bareja, Rasa Zarnegar, Brendan M Finnerty, Theresa Scognamiglio, Yi-Chieh Nancy Du, Olivier Elemento, Thomas J Fahey, Irene M Min
ABSTRACT
Pancreatic neuroendocrine tumors (PNETs) are malignancies arising from the islets of Langerhans. Therapeutic options are limited for the over 50% of patients who present with metastatic disease. We aimed to identify mechanisms to remodel the PNET tumor microenvironment (TME) to ultimately enhance susceptibility to immunotherapy. The TMEs of localized and metastatic PNETs were investigated using an approach that combines RNA-Seq, cancer and T cell profiling, and pharmacologic perturbations. RNA-Seq analysis indicated that the primary tumors of metastatic PNETs showed significant activation of inflammatory and immune-related pathways. We determined that metastatic PNETs featured increased numbers of tumor-infiltrating T cells compared with localized tumors. T cells isolated from both localized and metastatic PNETs showed evidence of recruitment and antigen-dependent activation, suggestive of an immune-permissive microenvironment. A computational analysis suggested that vorinostat, a histone deacetylase inhibitor, may perturb the transcriptomic signature of metastatic PNETs. Treatment of PNET cell lines with vorinostat increased chemokine CCR5 expression by NF-κB activation. Vorinostat treatment of patient-derived metastatic PNET tissues augmented recruitment of autologous T cells, and this augmentation was substantiated in a mouse model of PNET. Pharmacologic induction of chemokine expression may represent a promising approach for enhancing the immunogenicity of metastatic PNET TMEs.
Sigma-Aldrich
TPCA-1, ≥95% (HPLC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 mg
1 Estimated to ship on 24 February 2025
CHF 231.00
5 mg
1 Estimated to ship on 24 February 2025
CHF 665.00
Sigma-Aldrich
Human Serum, from human male AB plasma, USA origin, sterile-filtered
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 mL
1 Estimated to ship on 24 February 2025
CHF 116.00
100 mL
1 Estimated to ship on 24 February 2025
CHF 260.00
Sigma-Aldrich
2′,3′-cGAMP sodium salt, ≥98% (HPLC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
0.335 mg
1 Estimated to ship on 24 February 2025
CHF 317.00