Merck
  • TGF-β controls stromal telomere length through epigenetic modifications.
TGF-β controls stromal telomere length through epigenetic modifications.
3 Biotech (2022-10-25)
Rajeev Mishra, Subhash Haldar, Shea Biondi, Vikash Kumar Bhari, Gyanendra Singh, Neil A Bhowmick
ABSTRACT
Telomere length is primarily controlled by the enzyme telomerase, but being chromatin structures, telomeres undergo epigenetic regulation for their maintenance and function. Altered telomere length among cancer cells combined with shorter telomere length in cancer-associated stromal cells, strongly implicated with progression to prostate cancer metastasis and cancer death and providing a novel target for therapeutics. Transforming growth factor-β (TGF-β) signaling pathways are well-recognized for their role in stromal-epithelial interactions responsible for prostate androgen responsiveness, promoting tumorigenesis. However, the underlying mechanism remains unclear. We sought to establish a role for TGF-β in the regulation of telomere length in mouse and human prostate fibroblast. Polymerase chain reaction (PCR)-based telomere length measuring methods are widely used due to their repeatability and reproducibility. Using real-time RT-PCR-based telomere length measuring method, we identified that TGF-beta regulates telomere length via increased expression of histone methyltransferase, Suv39h1, which in turn affected histone methylation levels at the telomeric ends. Moreover, treatment of DAPT and non-steroidal antiandrogen bicalutamide demonstrated that notch and androgen signaling co-operated with TGF-ß in regulating stromal telomere length. Telomere variation in tumor cells and non-tumor cells within the tumor microenvironment greatly facilitates the clinical assessment of prostate cancer; therefore, understanding stromal telomere length regulation mechanism will hold significant prospects for cancer treatment, diagnosis, and prognosis. The online version contains supplementary material available at 10.1007/s13205-022-03346-5.
Sigma-Aldrich
Anti-HP1α Antibody, clone15.19s2, clone 15.19s2, Upstate®, from mouse