Merck
  • Proteome analysis of formalin-fixed paraffin-embedded colorectal adenomas reveals the heterogeneous nature of traditional serrated adenomas compared to other colorectal adenomas.
Proteome analysis of formalin-fixed paraffin-embedded colorectal adenomas reveals the heterogeneous nature of traditional serrated adenomas compared to other colorectal adenomas.
The Journal of pathology (2019-11-16)
Pierre Sohier, Romain Sanson, Marjorie Leduc, Anne Audebourg, Cédric Broussard, Virginie Salnot, Pierre-Alexandre Just, Eric Pasmant, Patrick Mayeux, François Guillonneau, Béatrice Romagnolo, Christine Perret, Benoît Terris
ABSTRACT
Traditional serrated adenoma (TSA) remains the least understood of all the colorectal adenomas, although these lesions have been associated with a significant cancer risk, twice that of the conventional adenoma (CAD) and of the sessile serrated adenoma (SSA/P). This study was performed to investigate the proteomic profiles of the different colorectal adenomas to better understand the pathogenesis of TSA. We performed a global quantitative proteome analysis using the label-free quantification (LFQ) method on 44 colorectal adenoma (12 TSAs, 15 CADs, and 17 SSA/Ps) and 17 normal colonic mucosa samples, archived as formalin-fixed paraffin-embedded blocks. Unsupervised consensus hierarchical clustering applied to the whole proteomic profile of the 44 colorectal adenomas identified four subtypes: C1 and C2 were well-individualized clusters composed of all the CADs (15/15) and most of the SSA/Ps (13/17), respectively. This is consistent with the fact that CADs and SSA/Ps are homogeneous and distinct colorectal adenoma entities. In contrast, TSAs were subdivided into C3 and C4 clusters, consistent with the more heterogeneous entity of TSA at the morphologic and molecular levels. Comparison of the proteome expression profile between the adenoma subtypes and normal colonic mucosa further confirmed the heterogeneous nature of TSAs, which overlapped either on CADs or SSA/Ps, whereas CADs and SSAs formed homogeneous and distinct entities. Furthermore, we identified LEFTY1 a new potential marker for TSAs that may be relevant for the pathogenesis of TSA. LEFTY1 is an inhibitor of the Nodal/TGFβ pathway, which we found to be one of the most overexpressed proteins specifically in TSAs. This finding was confirmed by immunohistochemistry. Our study confirms that CADs and SSA/Ps form homogeneous and distinct colorectal adenoma entities, whereas TSAs are a heterogeneous entity and may arise from either SSA/Ps or from normal mucosa evolving through a process related to the CAD pathway. © 2019 Pathological Society of Great Britain and Ireland. Published by John Wiley & Sons, Ltd.
Sigma-Aldrich
Anti-ANXA10 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 μL
Please contact Customer Service for Availability
₹58,263.90