Merck
  • Hypertension secondary to nitric oxide depletion produces oxidative imbalance and inflammatory/fibrotic outcomes in the cornea of C57BL/6 mice.
Hypertension secondary to nitric oxide depletion produces oxidative imbalance and inflammatory/fibrotic outcomes in the cornea of C57BL/6 mice.
Journal of physiology and biochemistry (2022-08-10)
Álvaro Santana-Garrido, Claudia Reyes-Goya, Ana Arroyo-Barrios, Helder André, Carmen M Vázquez, Alfonso Mate
ABSTRACT
Arterial hypertension (AH) leads to oxidative and inflammatory imbalance that contribute to fibrosis development in many target organs. Here, we aimed to highlight the harmful effects of severe AH in the cornea. Our experimental model was established by administration of NG-nitro-L-arginine-methyl-ester (L-NAME) to C57BL/6 mice, which were monitored weekly for arterial blood pressure and intraocular pressure (IOP). Morphological studies of ocular tissues were accompanied by analyses of reactive oxygen species generation, and localization/expression of NAPDH oxidase isoforms (NOX1, NOX2, NOX4) and inflammatory biomarkers (PPARα, PPARγ, IL-1β, IL-6, IL-10, TNF-α, and COX-2). Masson's trichrome and Sirius Red staining were used to explore the fibrotic status of the cornea. The expression of collagen isoforms (COL1α1, COL1α2, COL3α1, COL4α1, COL4α2) and relevant metalloproteinases (MMPs) and their tissue inhibitors (TIMPs) were also quantified to evaluate the participation of collagen metabolism in AH-related corneal damage. Hypertensive animals showed an increase in IOP values, and a thinner cornea compared with normotensive controls. Moreover, AH increased NADPH oxidase activity and reactive oxygen species generation in the cornea, which was accompanied by transcriptional upregulation of NOX isoforms and inflammatory biomarkers, while reducing PPAR expression. L-NAME-treated animals also developed corneal fibrosis with overexpression of collagen isoforms and reduction of factors responsible for collagen degradation. This is the first study reporting structural changes in the cornea and elevated IOP in L-NAME-treated mice. Overexpression of the NADPH oxidase system and collagen deposition might play a substantial role in the pathogenic mechanisms contributing to ocular disturbances in a context of severe hypertension.
Sigma-Aldrich
Masson-Goldner staining kit, for the visualization of connective tissue with trichromic staining
Sigma-Aldrich
Superoxide dismutase–polyethylene glycol from bovine erythrocytes, lyophilized powder, ≥1,350 units/mg protein