Merck
  • Biallelic loss-of-function variants in PLD1 cause congenital right-sided cardiac valve defects and neonatal cardiomyopathy.
Biallelic loss-of-function variants in PLD1 cause congenital right-sided cardiac valve defects and neonatal cardiomyopathy.
The Journal of clinical investigation (2021-03-02)
Najim Lahrouchi, Alex V Postma, Christian M Salazar, Daniel M De Laughter, Fleur Tjong, Lenka Piherová, Forrest Z Bowling, Dominic Zimmerman, Elisabeth M Lodder, Asaf Ta-Shma, Zeev Perles, Leander Beekman, Aho Ilgun, Quinn Gunst, Mariam Hababa, Doris Škorić-Milosavljević, Viktor Stránecký, Viktor Tomek, Peter de Knijff, Rick de Leeuw, Jamille Y Robinson, Sabrina C Burn, Hiba Mustafa, Matthew Ambrose, Timothy Moss, Jennifer Jacober, Dmitriy M Niyazov, Barry Wolf, Katherine H Kim, Sara Cherny, Andreas Rousounides, Aphrodite Aristidou-Kallika, George Tanteles, Bruel Ange-Line, Anne-Sophie Denommé-Pichon, Christine Francannet, Damara Ortiz, Monique C Haak, Arend D.J. Ten Harkel, Gwendolyn Tr Manten, Annemiek C Dutman, Katelijne Bouman, Monia Magliozzi, Francesca Clementina Radio, Gijs We Santen, Johanna C Herkert, H Alex Brown, Orly Elpeleg, Maurice Jb van den Hoff, Barbara Mulder, Michael V Airola, Stanislav Kmoch, Joey V Barnett, Sally-Ann Clur, Michael A Frohman, Connie R Bezzina
ABSTRACT
Congenital heart disease is the most common type of birth defect, accounting for one-third of all congenital anomalies. Using whole-exome sequencing of 2718 patients with congenital heart disease and a search in GeneMatcher, we identified 30 patients from 21 unrelated families of different ancestries with biallelic phospholipase D1 (PLD1) variants who presented predominantly with congenital cardiac valve defects. We also associated recessive PLD1 variants with isolated neonatal cardiomyopathy. Furthermore, we established that p.I668F is a founder variant among Ashkenazi Jews (allele frequency of ~2%) and describe the phenotypic spectrum of PLD1-associated congenital heart defects. PLD1 missense variants were overrepresented in regions of the protein critical for catalytic activity, and, correspondingly, we observed a strong reduction in enzymatic activity for most of the mutant proteins in an enzymatic assay. Finally, we demonstrate that PLD1 inhibition decreased endothelial-mesenchymal transition, an established pivotal early step in valvulogenesis. In conclusion, our study provides a more detailed understanding of disease mechanisms and phenotypic expression associated with PLD1 loss of function.
Sigma-Aldrich
VU0155056, ≥98% (HPLC)