Merck
  • Lead-Time Trajectory of CA19-9 as an Anchor Marker for Pancreatic Cancer Early Detection.
Lead-Time Trajectory of CA19-9 as an Anchor Marker for Pancreatic Cancer Early Detection.
Gastroenterology (2020-12-18)
Johannes F Fahrmann, C Max Schmidt, Xiangying Mao, Ehsan Irajizad, Maureen Loftus, Jinming Zhang, Nikul Patel, Jody Vykoukal, Jennifer B Dennison, James P Long, Kim-Anh Do, Jianjun Zhang, John A Chabot, Michael D Kluger, Fay Kastrinos, Lauren Brais, Ana Babic, Kunal Jajoo, Linda S Lee, Thomas E Clancy, Kimmie Ng, Andrea Bullock, Jeanine Genkinger, Michele T Yip-Schneider, Anirban Maitra, Brian M Wolpin, Samir Hanash
ABSTRACT
There is substantial interest in liquid biopsy approaches for cancer early detection among subjects at risk, using multi-marker panels. CA19-9 is an established circulating biomarker for pancreatic cancer; however, its relevance for pancreatic cancer early detection or for monitoring subjects at risk has not been established. CA19-9 levels were assessed in blinded sera from 175 subjects collected up to 5 years before diagnosis of pancreatic cancer and from 875 matched controls from the Prostate, Lung, Colorectal and Ovarian (PLCO) Cancer Screening Trial. For comparison of performance, CA19-9 was assayed in blinded independent sets of samples collected at diagnosis from 129 subjects with resectable pancreatic cancer and 275 controls (100 healthy subjects; 50 with chronic pancreatitis; and 125 with noncancerous pancreatic cysts). The complementary value of 2 additional protein markers, TIMP1 and LRG1, was determined. In the PLCO cohort, levels of CA19-9 increased exponentially starting at 2 years before diagnosis with sensitivities reaching 60% at 99% specificity within 0 to 6 months before diagnosis for all cases and 50% at 99% specificity for cases diagnosed with early-stage disease. Performance was comparable for distinguishing newly diagnosed cases with resectable pancreatic cancer from healthy controls (64% sensitivity at 99% specificity). Comparison of resectable pancreatic cancer cases to subjects with chronic pancreatitis yielded 46% sensitivity at 99% specificity and for subjects with noncancerous cysts, 30% sensitivity at 99% specificity. For prediagnostic cases below cutoff value for CA19-9, the combination with LRG1 and TIMP1 yielded an increment of 13.2% in sensitivity at 99% specificity (P = .031) in identifying cases diagnosed within 1 year of blood collection. CA19-9 can serve as an anchor marker for pancreatic cancer early detection applications.
Product Description
Millipore
MILLIPLEX® Human TIMP Magnetic Bead Panel 1 - Immunology Multiplex Assay, TIMP Bead-Based Multiplex Assays using the Luminex technology enables the simultaneous analysis of multiple TIMP biomarkers in human serum, plasma and cell culture samples.