Merck
  • Comparative analysis and generation of a robust HIV-1 DNA quantification assay.
Comparative analysis and generation of a robust HIV-1 DNA quantification assay.
Journal of virological methods (2018-10-17)
Jordan Thomas, Alessandra Ruggiero, Francesco A Procopio, Giuseppe Pantaleo, William A Paxton, Georgios Pollakis
ABSTRACT
HIV-1 infection cannot be cured due to the presence of the latent reservoir (LR). Novel cure or treatment strategies, such as "shock and kill" or therapeutic vaccination, aim to reduce or eradicate the LR. Cure strategies utilise robust DNA quantification assays to measure the change in the LR in low copy scenarios. No standard assay exists, which impedes the reliable comparison of results from different therapy and vaccine trials and HIV-1 total DNA quantification methods have not been previously compared. The HIV-1 long terminal repeat (LTR) has been shown to be the best target for DNA quantification. We have analysed two HIV-1 quantification assays, both able to differentiate between the variant HIV-1 DNA forms via the use of pre-amplification and primers targeting LTR. We identify a strong correlation (r=0.9759, P<0.0001) between assays which is conserved in low copy samples (r=0.8220, P<0.0001) indicating that these assays may be used interchangeably. The RvS assay performed significantly (P=0.0021) better than the CV assay when quantifying HIV-1 total DNA in patient CD4+ T lymphocytes. Sequence analysis demonstrated that viral diversity can limit DNA quantification, however in silico analysis of the primers indicated that within the target region nucleotide miss-matches appear infrequently. Further in silico analysis using up to-date sequence information led to the improvement of primers and enabled us to establish a more broadly specific assay with significantly higher HIV-1 DNA quantification capacity in patient samples (p=0.0057, n=17).
Sigma-Aldrich
Fetal Bovine Serum, non-USA origin, sterile-filtered, suitable for cell culture
Roche
Proteinase K, recombinant, PCR Grade, Lyophilizate from Pichia pastoris