Merck
  • Oligodendrocyte Progenitor Cell Proliferation and Fate after White Matter Stroke in Juvenile and Adult Mice.
Oligodendrocyte Progenitor Cell Proliferation and Fate after White Matter Stroke in Juvenile and Adult Mice.
Developmental neuroscience (2019-03-13)
Andra L Dingman, Krista M Rodgers, Robert M Dietz, Sean P Hickey, Alexandra P Frazier, Amy C Clevenger, Joan C Yonchek, Richard J Traystman, Wendy B Macklin, Paco S Herson
ABSTRACT
The incidence of stroke in children is 2.4 per 100,000 person-years and results in long-term motor and cognitive disability. In ischemic stroke, white matter (WM) is frequently injured, but is relatively understudied compared to grey matter injury. Previous research suggests that the cellular response to WM ischemic injury is different at different ages. Little is known about whether WM repair mechanisms differ in children and adults. We utilized a model of focal ischemic WM injury to determine the oligodendrocyte (OL) response to focal WM ischemic injury in juvenile and adult mice. Methods: Juvenile (21-25 days of age) versus adult (2-3 months of age) mice underwent stereotaxic injection of the potent vasoconstrictor N5-(1-iminoethyhl)-L-ornithine (L-NIO) into the lateral corpus callosum (CC). Animals were sacrificed on postoperative day 3 (acute) or 21 (chronic). Cell birth-dating was performed acutely after WM stroke with 5-ethynyl-2-deoxyuridine (EdU) injected intraperitoneally. Immunohistochemistry was performed, as well as stereology, to measure injury volume. The acute oligodendrocyte progenitor cell (OPC) proliferation and the chronic OL cell fate were determined with immunohistochemistry. Compound action potentials were measured in the CC at acute and chronic time points. Results: Acutely WM injury volume was smaller in juveniles. There was significantly greater OPC proliferation in juvenile animals (acute) compared to adults, but newly born OLs did not survive and mature into myelinating cells at chronic time points. In addition, juveniles did not have improved histological or functional recovery when compared to adults. Protecting newly born OPCs is a potential therapeutic target in children with ischemic stroke.
Sigma-Aldrich
Anti-Olig-2 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-APC (Ab-1) Mouse mAb (FE9), liquid, clone FE9, Calbiochem®