Merck
  • Depletion of mitochondrial methionine adenosyltransferase α1 triggers mitochondrial dysfunction in alcohol-associated liver disease.
Depletion of mitochondrial methionine adenosyltransferase α1 triggers mitochondrial dysfunction in alcohol-associated liver disease.
Nature communications (2022-01-30)
Lucía Barbier-Torres, Ben Murray, Jin Won Yang, Jiaohong Wang, Michitaka Matsuda, Aaron Robinson, Aleksandra Binek, Wei Fan, David Fernández-Ramos, Fernando Lopitz-Otsoa, Maria Luque-Urbano, Oscar Millet, Nirmala Mavila, Hui Peng, Komal Ramani, Roberta Gottlieb, Zhaoli Sun, Suthat Liangpunsakul, Ekihiro Seki, Jennifer E Van Eyk, Jose M Mato, Shelly C Lu
ABSTRACT
MATα1 catalyzes the synthesis of S-adenosylmethionine, the principal biological methyl donor. Lower MATα1 activity and mitochondrial dysfunction occur in alcohol-associated liver disease. Besides cytosol and nucleus, MATα1 also targets the mitochondria of hepatocytes to regulate their function. Here, we show that mitochondrial MATα1 is selectively depleted in alcohol-associated liver disease through a mechanism that involves the isomerase PIN1 and the kinase CK2. Alcohol activates CK2, which phosphorylates MATα1 at Ser114 facilitating interaction with PIN1, thereby inhibiting its mitochondrial localization. Blocking PIN1-MATα1 interaction increased mitochondrial MATα1 levels and protected against alcohol-induced mitochondrial dysfunction and fat accumulation. Normally, MATα1 interacts with mitochondrial proteins involved in TCA cycle, oxidative phosphorylation, and fatty acid β-oxidation. Preserving mitochondrial MATα1 content correlates with higher methylation and expression of mitochondrial proteins. Our study demonstrates a role of CK2 and PIN1 in reducing mitochondrial MATα1 content leading to mitochondrial dysfunction in alcohol-associated liver disease.
Sigma-Aldrich
Anti-Pin1 Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-β-Actin−Peroxidase antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture