Merck
  • Determination of pyrethrin and pyrethroid residues in animal fat using liquid chromatography coupled to tandem mass spectrometry.
Determination of pyrethrin and pyrethroid residues in animal fat using liquid chromatography coupled to tandem mass spectrometry.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2018-02-08)
M Moloney, S Tuck, A Ramkumar, A Furey, M Danaher
ABSTRACT
A method was developed for the confirmatory and quantitative analysis of one pyrethrin and 18 pyrethroid residues in animal fat. Fat was extracted was collected from adipose tissue melted in an oven at 65 °C for 2 h. Fat samples (1 g) were dispersed with deactivated Florisil® sorbent and extracted with MeCN. Sample extracts were purified by cold temperature precipitation at -30 °C for 4 h and further purified using dispersive solid-phase extraction (d-SPE) clean-up in tubes containing 500 mg of Z-SEP+ and 125 mg of PSA bonded silica. Purified samples were analysed by ultra-high performance liquid chromatography coupled to tandem mass spectrometry (UHPLC-MS/MS) detection. Chromatographic separation was carried out on a Acquity C8 BEH column, using a binary gradient separation comprising of mobile phase A, 5 mM ammonium formate in water:MeOH (80:20, v/v,) and mobile phase B, 5 mM ammonium formate in MeOH. The mass spectrometer was operated in the positive electrospray ionisation mode (ESI(+)). Validation was performed following the 2002/657/EC guidelines. Trueness ranged between 84% and 143% and precision ranged between 3.9% and 29%. The developed method is particularly advantageous because the sample preparation procedure does not require complex sample extraction equipment and uses less solvent compared to other sample preparation protocols.
Product Description
Supelco
Supel QuE, Z-Sep+ Bulk, pkg of 20 g
Supelco
Supel QuE, Z-Sep Tube, centrifuge tube volume 2 mL, pack of 100 ea
Supelco
Supel QuE, Z-Sep Tube, pk of 50, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, Z-Sep Bulk
Supelco
Supel QuE, Z-Sep+ Tube, centrifuge tube volume 2 mL, pack of 100 ea
Supelco
Supel QuE, Z-Sep+ with MgSO4 Tube, pk of 50, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, Z-Sep+ Tube, pk of 50, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Tralomethrin, mixture of stereoisomers, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Supel QuE, Z-Sep/C18 Tube, pk of 50, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, Z-Sep/C18 Tube, centrifuge tube volume 2 mL, pack of 100