Merck
  • Study of the degradation products of bromopropylate, chlordimeform, coumaphos, cymiazole, flumethrin and tau-fluvalinate in aqueous media.
Study of the degradation products of bromopropylate, chlordimeform, coumaphos, cymiazole, flumethrin and tau-fluvalinate in aqueous media.
Talanta (2008-10-31)
E Corta, A Bakkali, A Barranco, L A Berrueta, B Gallo, F Vicente, S Bogdanov
ABSTRACT
Degradation processes of bromopropylate, coumaphos, chlordimeform, cymiazole, flumethrin and fluvalinate in aqueous media have been studied by HPLC. Cymiazole is stable at any tested pH (1-11), while bromopropylate, flumethrin and coumaphos are unstable at basic pH and chlordimeform and fluvalinate in neutral and basic media. The main degradation products have been identified by GC-MS. The reaction rate constants and half-lives were calculated and the effect of co-solvents on the rate and product profile was studied.
Product Description
Supelco
Bromopropylate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland