Merck
  • HuR up-regulates cell surface PD-L1 via stabilizing CMTM6 transcript in cancer.
HuR up-regulates cell surface PD-L1 via stabilizing CMTM6 transcript in cancer.
Oncogene (2021-03-03)
Yanbin Liu, Xingzhi Li, Hui Zhang, Mingming Zhang, Yanli Wei
ABSTRACT
Despite the well-established role of CMTM6 in the stabilization of cell surface PD-L1 in cancer cells, the mechanisms underlying CMTM6 expression and regulation are still largely unknown. Here we unexpectedly find a strikingly positive correlation between CMTM6 and Hu-Antigen R (HuR) expression in most types of cancer. Mechanistically, we elucidate HuR stabilizes CMTM6 mRNA via direct association with AU-rich elements (AREs) in its 3'UTR and predominantly up-regulates CMTM6, which is readily abolished by HuR-specific inhibitor, MS-444. Phenotypically, we notice abundant cell surface PD-L1 in HuR-high cancer cells, which significantly inhibits immune activation of co-cultured T cells as indicated by IL-2 production. Treatment with MS-444 completely relieves immune suppression imposed by HuR-overexpression and further stimulates immune responses. Ectopic HuR accelerates allograft tumor progression in vivo, which is greatly compromised by simultaneous administration with MS-444. Our study uncovers a novel mechanism in control of CMTM6 and therefore PD-L1 expression, and suggests the potential of combining HuR inhibitor with PD-1/PD-L1 antibodies for cancer immunotherapy.
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC)
Sigma-Aldrich
Monoclonal Anti-GZMB antibody produced in mouse, clone CLB-GB11, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-CD107a-APC antibody produced in mouse, clone H4A3, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Lectin from Phaseolus vulgaris (red kidney bean), Leucoagglutinin PHA-L, lyophilized powder
Sigma-Aldrich
Anti-CMTM6 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution