Merck
  • Discovery of N-(Naphthalen-1-yl)-N'-alkyl Oxalamide Ligands Enables Cu-Catalyzed Aryl Amination with High Turnovers.
Discovery of N-(Naphthalen-1-yl)-N'-alkyl Oxalamide Ligands Enables Cu-Catalyzed Aryl Amination with High Turnovers.
Organic letters (2017-05-23)
Jie Gao, Subhajit Bhunia, Kailiang Wang, Lu Gan, Shanghua Xia, Dawei Ma
ABSTRACT
A class of N-(naphthalen-1-yl)-N'-alkyl oxalamides have been proven to be powerful ligands, making a coupling reaction of (hetero)aryl iodides with primary amines proceed at 50 °C with only 0.01 mol % of Cu2O and ligand as well as a coupling reaction of (hetero)aryl bromides with primary amines and ammonia at 80 °C with only 0.1 mol % of Cu2O and ligand. A wide range of coupling partners work well under these conditions, thereby providing an easy to operate method for preparing (hetero)aryl amines.
Sigma-Aldrich
MNBO, ≥95%