Merck
  • Neurons for hunger and thirst transmit a negative-valence teaching signal.
Neurons for hunger and thirst transmit a negative-valence teaching signal.
Nature (2015-04-29)
J Nicholas Betley, Shengjin Xu, Zhen Fang Huang Cao, Rong Gong, Christopher J Magnus, Yang Yu, Scott M Sternson
ABSTRACT
Homeostasis is a biological principle for regulation of essential physiological parameters within a set range. Behavioural responses due to deviation from homeostasis are critical for survival, but motivational processes engaged by physiological need states are incompletely understood. We examined motivational characteristics of two separate neuron populations that regulate energy and fluid homeostasis by using cell-type-specific activity manipulations in mice. We found that starvation-sensitive AGRP neurons exhibit properties consistent with a negative-valence teaching signal. Mice avoided activation of AGRP neurons, indicating that AGRP neuron activity has negative valence. AGRP neuron inhibition conditioned preference for flavours and places. Correspondingly, deep-brain calcium imaging revealed that AGRP neuron activity rapidly reduced in response to food-related cues. Complementary experiments activating thirst-promoting neurons also conditioned avoidance. Therefore, these need-sensing neurons condition preference for environmental cues associated with nutrient or water ingestion, which is learned through reduction of negative-valence signals during restoration of homeostasis.
Sigma-Aldrich
PSEM89S Hydrochloride salt, ≥98% (HPLC)