Merck
Asymmetric Total Synthesis of C9'-epi-Sinefungin.
Organic letters (2020-07-07)
Ludovic Decultot, Rocco L Policarpo, Brandon A Wright, Danny Huang, Matthew D Shair
ABSTRACT
The natural nucleoside (+)-sinefungin, structurally similar to cofactor S-adenosyl-l-methionine, inhibits various SAM-dependent methyltransferases (MTs). Access to sinefungin analogues could serve as the basis for the rational design of small molecule methyltransferase inhibitors. We developed a route to the unnatural C9' epimer of sinefungin that employed a diastereoselective Overman rearrangement to install the key C6' amino stereocenter. The ability for late-stage modification is highlighted, opening an avenue for the discovery of new MT inhibitors.