Merck
  • Applying real-time monitoring of circadian oscillations in adult mouse brain slices to study communications between brain regions.
Applying real-time monitoring of circadian oscillations in adult mouse brain slices to study communications between brain regions.
STAR protocols (2021-04-20)
Suihong Huang, Qingqing Lu, Ming Ho Choi, Xuebing Zhang, Jin Young Kim
ABSTRACT
This protocol combines a protective cutting method to prepare various brain slices from adult mice and real-time monitoring of circadian oscillations to measure circadian rhythmicity in various brain slices. This protocol can be applied to studies of how brain damages affect local circadian clocks and subsequent circadian variations in nearby areas. Further functional analyses with in vivo systems will determine whether these circadian variations are detrimental or beneficial to the brain. For complete details on the use and execution of this protocol, please refer to Huang et al. (2020).
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - low glucose, With 1000 mg/L glucose and L-glutamine, without sodium bicarbonate and phenol red, powder, suitable for cell culture
Sigma-Aldrich
Hanks′ Balanced Salt Solution 10x, Without sodium bicarbonate, 10 ×, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Sodium bicarbonate solution, solution (7.5%), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium phosphate monobasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
L-α-Lysophosphatidylcholine from egg yolk, ≥99%, Type I, powder
Sigma-Aldrich
Choline chloride, ≥98%
Sigma-Aldrich
HEPES solution, 1 M, pH 7.0-7.6, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Lipopolysaccharides from Salmonella typhosa, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%