Merck
  • Functional Selectivity in Cytokine Signaling Revealed Through a Pathogenic EPO Mutation.
Functional Selectivity in Cytokine Signaling Revealed Through a Pathogenic EPO Mutation.
Cell (2017-03-12)
Ah Ram Kim, Jacob C Ulirsch, Stephan Wilmes, Ekrem Unal, Ignacio Moraga, Musa Karakukcu, Daniel Yuan, Shideh Kazerounian, Nour J Abdulhay, David S King, Namrata Gupta, Stacey B Gabriel, Eric S Lander, Turkan Patiroglu, Alper Ozcan, Mehmet Akif Ozdemir, K Christopher Garcia, Jacob Piehler, Hanna T Gazda, Daryl E Klein, Vijay G Sankaran
ABSTRACT
Cytokines are classically thought to stimulate downstream signaling pathways through monotonic activation of receptors. We describe a severe anemia resulting from a homozygous mutation (R150Q) in the cytokine erythropoietin (EPO). Surprisingly, the EPO R150Q mutant shows only a mild reduction in affinity for its receptor but has altered binding kinetics. The EPO mutant is less effective at stimulating erythroid cell proliferation and differentiation, even at maximally potent concentrations. While the EPO mutant can stimulate effectors such as STAT5 to a similar extent as the wild-type ligand, there is reduced JAK2-mediated phosphorylation of select downstream targets. This impairment in downstream signaling mechanistically arises from altered receptor dimerization dynamics due to extracellular binding changes. These results demonstrate how variation in a single cytokine can lead to biased downstream signaling and can thereby cause human disease. Moreover, we have defined a distinct treatable form of anemia through mutation identification and functional studies.
Sigma-Aldrich
CHZ868, ≥98% (HPLC)
Sigma-Aldrich
Giemsa Stain, Modified