Merck
  • POGLUT2 and POGLUT3 O-glucosylate multiple EGF repeats in fibrillin-1, -2, and LTBP1 and promote secretion of fibrillin-1.
POGLUT2 and POGLUT3 O-glucosylate multiple EGF repeats in fibrillin-1, -2, and LTBP1 and promote secretion of fibrillin-1.
The Journal of biological chemistry (2021-08-20)
Daniel B Williamson, Camron J Sohn, Atsuko Ito, Robert S Haltiwanger
ABSTRACT
Fibrillin-1 (FBN1) is the major component of extracellular matrix microfibrils, which are required for proper development of elastic tissues, including the heart and lungs. Through protein-protein interactions with latent transforming growth factor (TGF) β-binding protein 1 (LTBP1), microfibrils regulate TGF-β signaling. Mutations within the 47 epidermal growth factor-like (EGF) repeats of FBN1 cause autosomal dominant disorders including Marfan Syndrome, which is characterized by disrupted TGF-β signaling. We recently identified two novel protein O-glucosyltransferases, Protein O-glucosyltransferase 2 (POGLUT2) and 3 (POGLUT3), that modify a small fraction of EGF repeats on Notch. Here, using mass spectral analysis, we show that POGLUT2 and POGLUT3 also modify over half of the EGF repeats on FBN1, fibrillin-2 (FBN2), and LTBP1. While most sites are modified by both enzymes, some sites show a preference for either POGLUT2 or POGLUT3. POGLUT2 and POGLUT3 are homologs of POGLUT1, which stabilizes Notch proteins by addition of O-glucose to Notch EGF repeats. Like POGLUT1, POGLUT2 and 3 can discern a folded versus unfolded EGF repeat, suggesting POGLUT2 and 3 are involved in a protein folding pathway. In vitro secretion assays using the N-terminal portion of recombinant FBN1 revealed reduced FBN1 secretion in POGLUT2 knockout, POGLUT3 knockout, and POGLUT2 and 3 double-knockout HEK293T cells compared with wild type. These results illustrate that POGLUT2 and 3 function together to O-glucosylate protein substrates and that these modifications play a role in the secretion of substrate proteins. It will be interesting to see how disease variants in these proteins affect their O-glucosylation.
Sigma-Aldrich
Endoproteinase Glu-C from Staphylococcus aureus V8, suitable for protein sequencing, lyophilized powder