Merck
  • Augmented antibody-based anticancer therapeutics boost neutrophil cytotoxicity.
Augmented antibody-based anticancer therapeutics boost neutrophil cytotoxicity.
The Journal of clinical investigation (2021-02-10)
Niels Heemskerk, Mandy Gruijs, A Robin Temming, Marieke H Heineke, Dennis Y Gout, Tessa Hellingman, Cornelis W Tuk, Paula J Winter, Suzanne Lissenberg-Thunnissen, Arthur Eh Bentlage, Marco de Donatis, Marijn Bögels, Thies Rösner, Thomas Valerius, Jantine E Bakema, Gestur Vidarsson, Marjolein van Egmond
ABSTRACT
Most clinically used anticancer mAbs are of the IgG isotype, which can eliminate tumor cells through NK cell-mediated antibody-dependent cellular cytotoxicity and macrophage-mediated antibody-dependent phagocytosis. IgG, however, ineffectively recruits neutrophils as effector cells. IgA mAbs induce migration and activation of neutrophils through the IgA Fc receptor (FcαRI) but are unable to activate NK cells and have poorer half-life. Here, we combined the agonistic activity of IgG mAbs and FcαRI targeting in a therapeutic bispecific antibody format. The resulting TrisomAb molecules recruited NK cells, macrophages, and neutrophils as effector cells for eradication of tumor cells in vitro and in vivo. Moreover, TrisomAb had long in vivo half-life and strongly decreased B16F10gp75 tumor outgrowth in mice. Importantly, neutrophils of colorectal cancer patients effectively eliminated tumor cells in the presence of anti-EGFR TrisomAb but were less efficient in mediating killing in the presence of IgG anti-EGFR mAb (cetuximab). The clinical application of TrisomAb may provide potential alternatives for cancer patients who do not benefit from current IgG mAb therapy.
Sigma-Aldrich
Anti-Lactoferrin antibody produced in rabbit, fractionated antiserum, lyophilized powder
Sigma-Aldrich
SIGMAFAST p-Nitrophenyl phosphate Tablets, tablet, To prepare 5 mL
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Monoclonal Anti-Human IgG1 (Fab specific) [G1m(f)] antibody produced in mouse, clone SG-16, ascites fluid