Merck
  • NKD2 mediates stimulation-dependent ORAI1 trafficking to augment Ca2+ entry in T cells.
NKD2 mediates stimulation-dependent ORAI1 trafficking to augment Ca2+ entry in T cells.
Cell reports (2021-08-26)
Beibei Wu, Jin Seok Woo, Pamela Vila, Marcus Jew, Jennifer Leung, Zuoming Sun, Sonal Srikanth, Yousang Gwack
ABSTRACT
Sustained activation of the Ca2+-release-activated Ca2+ (CRAC) channel is pivotal for effector T cell responses. The mechanisms underlying this sustainability remain poorly understood. We find that plasma membrane localization of ORAI1, the pore subunit of CRAC channels, is limited in effector T cells, with a significant fraction trapped in intracellular vesicles. From a targeted screen, we identify an essential component of ORAI1+ vesicles, naked cuticle homolog 2 (NKD2). Mechanistically, NKD2, an adaptor molecule activated by signaling pathways downstream of T cell receptors, orchestrates trafficking and insertion of ORAI1+ vesicles to the plasma membrane. Together, our findings suggest that T cell receptor (TCR)-stimulation-dependent insertion of ORAI1 into the plasma membrane is essential for sustained Ca2+ signaling and cytokine production in T cells.
Product Description
Millipore
Ionomycin, Free Acid, Streptomyces conglobatus in Solution
Sigma-Aldrich
Polybrene Infection / Transfection Reagent, A highly efficient method of gene transfer into mammalian cells leveraging infection with retroviral vectors.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M1 antibody produced in mouse, clone M1, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-Orai1 Antibody, Intracellular, Chemicon®, from rabbit