Merck
  • Loss of lysosome-associated membrane protein 3 (LAMP3) enhances cellular vulnerability against proteasomal inhibition.
Loss of lysosome-associated membrane protein 3 (LAMP3) enhances cellular vulnerability against proteasomal inhibition.
European journal of cell biology (2015-02-15)
Jorge Antolio Dominguez-Bautista, Michael Klinkenberg, Nadine Brehm, Mahalakshmi Subramaniam, Beatrice Kern, Jochen Roeper, Georg Auburger, Marina Jendrach
ABSTRACT
The family of lysosome-associated membrane proteins (LAMP) includes the ubiquitously expressed LAMP1 and LAMP2, which account for half of the proteins in the lysosomal membrane. Another member of the LAMP family is LAMP3, which is expressed only in certain cell types and differentiation stages. LAMP3 expression is linked with poor prognosis of certain cancers, and the locus where it is encoded was identified as a risk factor for Parkinson's disease (PD). Here, we investigated the role of LAMP3 in the two main cellular degradation pathways, the proteasome and autophagy. LAMP3 mRNA was not detected in mouse models of PD or in the brain of human patients. However, it was strongly induced upon proteasomal inhibition in the neuroblastoma cell line SH-SY5Y. Induction of LAMP3 mRNA following proteasomal inhibition was dependent on UPR transcription factor ATF4 signaling and induced autophagic flux. Prevention of LAMP3 induction enhanced apoptotic cell death. In summary, these data demonstrate that LAMP3 regulation as part of the UPR contributes to protein degradation and cell survival during proteasomal dysfunction. This link between autophagy and the proteasome may be of special importance for the treatment of tumor cells with proteasomal inhibitors.
Sigma-Aldrich
Anti-LAMP-3 Antibody, clone 16H11.2, clone 16H11.2, from mouse