Merck
  • Splice site m6A methylation prevents binding of U2AF35 to inhibit RNA splicing.
Splice site m6A methylation prevents binding of U2AF35 to inhibit RNA splicing.
Cell (2021-05-01)
Mateusz Mendel, Kamila Delaney, Radha Raman Pandey, Kuan-Ming Chen, Joanna M Wenda, Cathrine Broberg Vågbø, Florian A Steiner, David Homolka, Ramesh S Pillai
ABSTRACT
The N6-methyladenosine (m6A) RNA modification is used widely to alter the fate of mRNAs. Here we demonstrate that the C. elegans writer METT-10 (the ortholog of mouse METTL16) deposits an m6A mark on the 3' splice site (AG) of the S-adenosylmethionine (SAM) synthetase pre-mRNA, which inhibits its proper splicing and protein production. The mechanism is triggered by a rich diet and acts as an m6A-mediated switch to stop SAM production and regulate its homeostasis. Although the mammalian SAM synthetase pre-mRNA is not regulated via this mechanism, we show that splicing inhibition by 3' splice site m6A is conserved in mammals. The modification functions by physically preventing the essential splicing factor U2AF35 from recognizing the 3' splice site. We propose that use of splice-site m6A is an ancient mechanism for splicing regulation.
Sigma-Aldrich
(1R,2S,5R)-(−)-Menthol, ReagentPlus®, 99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Vitamin B12, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ribonuclease A from bovine pancreas, for molecular biology, ≥70 Kunitz units/mg protein, lyophilized
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Nuclease P1 from Penicillium citrinum, lyophilized powder, ≥200 units/mg protein (E1%/280, 3′-5′-Phosphodiesterase)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
TWEEN® 20, BioXtra, viscous liquid
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Cystine, from non-animal source, meets EP testing specifications, suitable for cell culture, 98.5-101.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Leucine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Eosin Y disodium salt, certified by the Biological Stain Commission
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Bouin′s solution, histological fixative
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Methionine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Folic acid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥97%