Merck
A murine closed-chest model of myocardial ischemia and reperfusion.
Journal of visualized experiments : JoVE (2012-08-01)
Se-Chan Kim, Olaf Boehm, Rainer Meyer, Andreas Hoeft, Pascal Knüfermann, Georg Baumgarten
ABSTRACT
Surgical trauma by thoracotomy in open-chest models of coronary ligation induces an immune response which modifies different mechanisms involved in ischemia and reperfusion. Immune response includes cytokine expression and release or secretion of endogenous ligands of innate immune receptors. Activation of innate immunity can potentially modulate infarct size. We have modified an existing murine closed-chest model using hanging weights which could be useful for studying myocardial pre- and postconditioning and the role of innate immunity in myocardial ischemia and reperfusion. This model allows animals to recover from surgical trauma before onset of myocardial ischemia. Volatile anesthetics have been intensely studied and their preconditioning effect for the ischemic heart is well known. However, this protective effect precludes its use in open chest models of coronary artery ligation. Thus, another advantage could be the use of the well controllable volatile anesthetics for instrumentation in a chronic closed-chest model, since their preconditioning effect lasts up to 72 hours. Chronic heart diseases with intermittent ischemia and multiple hit models are other possible applications of this model. For the chronic closed-chest model, intubated and ventilated mice undergo a lateral blunt thoracotomy via the 4th intercostal space. Following identification of the left anterior descending a ligature is passed underneath the vessel and both suture ends are threaded through an occluder. Then, both suture ends are passed through the chest wall, knotted to form a loop and left in the subcutaneous tissue. After chest closure and recovery for 5 days, mice are anesthetized again, chest skin is reopened and hanging weights are hooked up to the loop under ECG control. At the end of the ischemia/reperfusion protocol, hearts can be stained with TTC for infarct size assessment or undergo perfusion fixation to allow morphometric studies in addition to histology and immunohistochemistry.
Sigma-Aldrich
1,3,5-Triphenyltetrazolium formazan, ≥90% (UV)