Merck
  • TRIB2 desensitizes ferroptosis via βTrCP-mediated TFRC ubiquitiantion in liver cancer cells.
TRIB2 desensitizes ferroptosis via βTrCP-mediated TFRC ubiquitiantion in liver cancer cells.
Cell death discovery (2021-07-29)
Susu Guo, Yuxin Chen, Xiangfei Xue, Yueyue Yang, Yikun Wang, Shiyu Qiu, Jiangtao Cui, Xiao Zhang, Lifang Ma, Yongxia Qiao, Jiayi Wang
ABSTRACT
Tribbles homolog 2 (TRIB2) is known to boost liver tumorigenesis via regulating Ubiquitin (Ub) proteasome system (UPS). At least two ways are involved, i.e., acts as an adaptor protein to modulate ubiquitination functions of certain ubiquitin E3 ligases (E3s) and reduces global Ub levels via increasing the proteolysis activity of proteasome. Recently, we have identified the role of TRIB2 to relieve oxidative damage via reducing the availability of Ub that is essential for the ubiquitination and subsequent degradation of Glutathione peroxidase 4 (GPX4). Although GPX4 is a critical antioxidant factor to protect against ferroptosis, the exact evidence showing that TRIB2 desensitizes ferroptosis is lacking. Also, whether such function is via E3 remains unclear. Here, we demonstrated that deletion of TRIB2 sensitized ferroptosis via lifting labile iron in liver cancer cells. By contrast, overexpression of TRIB2 led to the opposite outcome. We further demonstrated that transferrin receptor (TFRC) was required for TRIB2 to desensitize the cells to ferroptosis. Without TFRC, the labile iron pool could not be reduced by overexpressing TRIB2. We also found that beta-transducin repeat containing E3 ubiqutin protein ligase (βTrCP) was a genuine E3 for the ubiquitination of TFRC, and TRIB2 was unable to decline labile iron level once upon βTrCP was knocked out. In addition, we confirmed that the opposite effects on ferroptosis and ferroptosis-associated lipid reactive oxygen species (ROS) generation resulted from knockout and overexpression of TRIB2 were all indispensible of TFRC and βTrCP. Finally, we demonstrated that TRIB2 exclusively manipulated RSL3- and erastin-induced-ferroptosis independent of GPX4 and glutathione (GSH). In conclusion, we elucidated a novel role of TRIB2 to desensitize ferroptosis via E3 βTrCP, by which facilitates TFRC ubiquitiation and finally decreases labile iron in liver cancer cells.
Sigma-Aldrich
Fatty Acid Extraction Kit, Low Standard, sufficient for 40 extractions
Sigma-Aldrich
Deferoxamine mesylate salt, powder, ≥92.5% (TLC)